N-(tert-Butyl)-6-chloro-4-pyrimidinamine

Supplier: Matrix Scientific

052890-500MG 052890-2.5G
101901-448EA 364.62 USD
101901-448 101901-450
N-(tert-Butyl)-6-chloro-4-pyrimidinamine
N-(tert-Butyl)-6-chloro-4-pyrimidinamine

Hazard: Irritant

Formula: C₈H₁₂ClN₃
MW: 185.66 g/mol
MDL Number: MFCD09756959
CAS Number: 945896-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR