N-(5-Bromo-4-methyl-2-pyridinyl)-N-(tert-butyl)amine

Supplier: Matrix Scientific

052889-500MG 052889-2.5G
101901-444EA 364.62 USD
101901-444 101901-446
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(tert-butyl)amine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(tert-butyl)amine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂
MDL Number: MFCD13562433
CAS Number: 1220018-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR