4-(tert-Butylamino)-3-nitrobenzoic acid

Supplier: Matrix Scientific

052888-500MG 052888-2.5G
101901-440EA 364.62 USD
101901-440 101901-442
4-(tert-Butylamino)-3-nitrobenzoic acid
4-(tert-Butylamino)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₄
MDL Number: MFCD09031717
CAS Number: 691363-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR