Ethyl-3-amino-4-(tert-butylamino)benzoate

Supplier: Matrix Scientific

052887-500MG 052887-2.5G
101901-436EA 364.62 USD
101901-436 101901-438
Ethyl-3-amino-4-(tert-butylamino)benzoate
Ethyl-3-amino-4-(tert-butylamino)benzoate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MDL Number: MFCD13343591
CAS Number: 637041-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR