3-Amino-4-chloro-N-(2-hydroxyethyl)benzenesulfonamide

Supplier: Matrix Scientific

052886-500MG 052886-2.5G
101901-432EA 364.62 USD
101901-432 101901-434
3-Amino-4-chloro-N-(2-hydroxyethyl)benzenesulfonamide
3-Amino-4-chloro-N-(2-hydroxyethyl)benzenesulfonamide

Hazard: Irritant

Formula: C₈H₁₁ClN₂O₃S
MW: 250.71 g/mol
MDL Number: MFCD11642919
CAS Number: 1098363-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR