N-(2-Hydroxyethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Supplier: Matrix Scientific
052885-500MG
052885-2.5G
101901-428EA
364.62
USD
101901-428
101901-430
N-(2-Hydroxyethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-(2-Hydroxyethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₅ClN₄O₂ MW: 246.7 g/mol |
MDL Number:
MFCD13562432 CAS Number: 1220035-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...