N-(2-Hydroxyethyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052884-500MG 052884-2.5G
101901-424EA 364.62 USD
101901-424 101901-426
N-(2-Hydroxyethyl)-4-piperidinecarboxamide hydrochloride
N-(2-Hydroxyethyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O₂
MDL Number: MFCD11615739
CAS Number: 30672-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR