N-(2-Hydroxyethyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052882-500MG 052882-2.5G
101901-416EA 364.62 USD
101901-416 101901-418
N-(2-Hydroxyethyl)-2-piperidinecarboxamide hydrochloride
N-(2-Hydroxyethyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O₂
MDL Number: MFCD13562430
CAS Number: 1236262-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR