2-Amino-N-(2-hydroxyethyl)-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052880-500MG 052880-2.5G
101901-408EA 364.62 USD
101901-408 101901-410
2-Amino-N-(2-hydroxyethyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(2-hydroxyethyl)-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O₂
MW: 244.72 g/mol
MDL Number: MFCD13562428
CAS Number: 1246172-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR