2-Amino-N-(2-hydroxyethyl)-2-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052879-500MG 052879-2.5G
101901-404EA 364.62 USD
101901-404 101901-406
2-Amino-N-(2-hydroxyethyl)-2-methylpropanamide hydrochloride
2-Amino-N-(2-hydroxyethyl)-2-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂
MDL Number: MFCD13562427
CAS Number: 1220034-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR