2-Amino-N-(2-hydroxyethyl)-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052878-500MG 052878-2.5G
101901-400EA 364.62 USD
101901-400 101901-402
2-Amino-N-(2-hydroxyethyl)-3-methylbutanamide hydrochloride
2-Amino-N-(2-hydroxyethyl)-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂
MDL Number: MFCD13562426
CAS Number: 1236263-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR