3-Amino-N-(2-hydroxyethyl)propanamide hydrochloride

Supplier: Matrix Scientific

052877-500MG 052877-2.5G
101901-396EA 364.62 USD
101901-396 101901-398
3-Amino-N-(2-hydroxyethyl)propanamide hydrochloride
3-Amino-N-(2-hydroxyethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O₂
MDL Number: MFCD13562425
CAS Number: 1220037-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR