N-(2-Hydroxyethyl)-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052876-500MG 052876-2.5G
101901-392EA 364.62 USD
101901-392 101901-394
N-(2-Hydroxyethyl)-2-(methylamino)acetamide hydrochloride
N-(2-Hydroxyethyl)-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O₂
MDL Number: MFCD13562424
CAS Number: 1220029-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR