2-Amino-N-(2-hydroxyethyl)acetamide hydrochloride

Supplier: Matrix Scientific

052874-500MG 052874-2.5G
101901-384EA 213.85 USD
101901-384 101901-386
2-Amino-N-(2-hydroxyethyl)acetamide hydrochloride
2-Amino-N-(2-hydroxyethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂O₂
MDL Number: MFCD13562422
CAS Number: 86150-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR