2-[(6-Chloro-2-pyridinyl)amino]-1-ethanol

Supplier: Matrix Scientific

Synonyms: 2-((6-Chloropyridin-2-yl)amino)ethanol

052873-500MG 052873-2.5G
101901-380EA 364.62 USD
101901-380 101901-382
2-[(6-Chloro-2-pyridinyl)amino]-1-ethanol
2-[(6-Chloro-2-pyridinyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₇H₉ClN₂O
MW: 172.62 g/mol
MDL Number: MFCD13562421
CAS Number: 29449-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR