2-[(5-Bromo-2-pyridinyl)amino]-1-ethanol
Supplier: Matrix Scientific
052872-500MG
052872-2.5G
101901-376EA
364.62
USD
101901-376
101901-378
2-[(5-Bromo-2-pyridinyl)amino]-1-ethanol
2-[(5-Bromo-2-pyridinyl)amino]-1-ethanol
Hazard: Irritant
Formula:
C₇H₉BrN₂O |
MDL Number:
MFCD11123700 CAS Number: 911112-04-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...