2-[(5-Bromo-3-methyl-2-pyridinyl)amino]-1-ethanol

Supplier: Matrix Scientific

052871-500MG 052871-2.5G
101901-372EA 364.62 USD
101901-372 101901-374
2-[(5-Bromo-3-methyl-2-pyridinyl)amino]-1-ethanol
2-[(5-Bromo-3-methyl-2-pyridinyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₈H₁₁BrN₂O
MDL Number: MFCD13562420
CAS Number: 1219982-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR