2-[(3-Amino-2-pyridinyl)amino]-1-ethanol

Supplier: Matrix Scientific

052869-500MG 052869-2.5G
101901-364EA 166.15 USD
101901-364 101901-366
2-[(3-Amino-2-pyridinyl)amino]-1-ethanol
2-[(3-Amino-2-pyridinyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₇H₁₁N₃O
MDL Number: MFCD11101081
CAS Number: 118705-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR