2-[(2-Hydroxyethyl)amino]nicotinic acid

Supplier: Matrix Scientific

052868-500MG 052868-2.5G
101901-360EA 364.62 USD
101901-360 101901-362
2-[(2-Hydroxyethyl)amino]nicotinic acid
2-[(2-Hydroxyethyl)amino]nicotinic acid

Hazard: Irritant

Formula: C₈H₁₀N₂O₃
MDL Number: MFCD13562419
CAS Number: 1220018-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR