2-[(2-Hydroxyethyl)amino]isonicotinic acid

Supplier: Matrix Scientific

052867-500MG 052867-2.5G
101901-356EA 384.62 USD
101901-356 101901-358
2-[(2-Hydroxyethyl)amino]isonicotinic acid
2-[(2-Hydroxyethyl)amino]isonicotinic acid

Hazard: Irritant

Formula: C₈H₁₀N₂O₃
MW: 182.18 g/mol
MDL Number: MFCD13562418
CAS Number: 1220018-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR