2-[(6-Chloro-4-pyrimidinyl)amino]-1-ethanol

Supplier: Matrix Scientific

Synonyms: 2-((6-Chloropyrimidin-4-yl)amino)ethanol

052864-500MG 052864-2.5G
101901-344EA 364.62 USD
101901-344 101901-346
2-[(6-Chloro-4-pyrimidinyl)amino]-1-ethanol
2-[(6-Chloro-4-pyrimidinyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₆H₈ClN₃O
MW: 173.6 g/mol
MDL Number: MFCD03672827
CAS Number: 22177-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR