2-[(5-Bromo-4-methyl-2-pyridinyl)amino]-1-ethanol

Supplier: Matrix Scientific

Synonyms: 2-((5-Bromo-4-methylpyridin-2-yl)amino)ethanol

052863-500MG 052863-2.5G
101901-340EA 384.62 USD
101901-340 101901-342
2-[(5-Bromo-4-methyl-2-pyridinyl)amino]-1-ethanol
2-[(5-Bromo-4-methyl-2-pyridinyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₈H₁₁BrN₂O
MW: 231.1 g/mol
MDL Number: MFCD13562416
CAS Number: 1219982-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR