4-[(2-Hydroxyethyl)amino]-3-nitrobenzoic acid

Supplier: Matrix Scientific

052862-500MG 052862-2.5G
101901-336EA 364.62 USD
101901-336 101901-338
4-[(2-Hydroxyethyl)amino]-3-nitrobenzoic acid
4-[(2-Hydroxyethyl)amino]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₉H₁₀N₂O₅
MDL Number: MFCD00227244
CAS Number: 59320-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR