Ethyl-3-amino-4-[(2-hydroxyethyl)amino]benzoate

Supplier: Matrix Scientific

052861-500MG 052861-2.5G
101901-332EA 364.62 USD
101901-332 101901-334
Ethyl-3-amino-4-[(2-hydroxyethyl)amino]benzoate
Ethyl-3-amino-4-[(2-hydroxyethyl)amino]benzoate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃
MDL Number: MFCD13562415
CAS Number: 91430-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR