N-Isopropyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052858-500MG 052858-2.5G
101901-320EA 384.62 USD
101901-320 101901-322
N-Isopropyl-4-piperidinecarboxamide hydrochloride
N-Isopropyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD11615741
CAS Number: 102125-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR