N-Isopropyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052857-500MG 052857-2.5G
101901-316EA 364.62 USD
101901-316 101901-318
N-Isopropyl-3-piperidinecarboxamide hydrochloride
N-Isopropyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562413
CAS Number: 937725-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR