N-Isopropyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052856-500MG 052856-2.5G
101901-312EA 364.62 USD
101901-312 101901-314
N-Isopropyl-2-piperidinecarboxamide hydrochloride
N-Isopropyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562412
CAS Number: 1236261-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR