2-Amino-N-isopropyl-2-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052853-500MG 052853-2.5G
101901-300EA 364.62 USD
101901-300 101901-302
2-Amino-N-isopropyl-2-methylpropanamide hydrochloride
2-Amino-N-isopropyl-2-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD13562409
CAS Number: 1220027-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR