2-Amino-N-isopropyl-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052852-500MG 052852-2.5G
101901-296EA 364.62 USD
101901-296 101901-298
2-Amino-N-isopropyl-3-methylbutanamide hydrochloride
2-Amino-N-isopropyl-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₉ClN₂O
MDL Number: MFCD13562408
CAS Number: 1236256-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR