1-[4-(3-Pyrrolidinylmethoxy)phenyl]-1-ethanone hydrochloride

Supplier: Matrix Scientific

049787-500MG 049787-2.5G
101889-048EA 364.62 USD
101889-048 101889-050
1-[4-(3-Pyrrolidinylmethoxy)phenyl]-1-ethanone hydrochloride
1-[4-(3-Pyrrolidinylmethoxy)phenyl]-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₂
MDL Number: MFCD13560189
CAS Number: 1220029-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR