1-{4-[2-(2-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride
Supplier: Matrix Scientific
049786-500MG
049786-2.5G
101889-044EA
364.62
USD
101889-044
101889-046
1-{4-[2-(2-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride
1-{4-[2-(2-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₂ClNO₂ |
MDL Number:
MFCD13560188 CAS Number: 1220031-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...