1-{4-[2-(3-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride
Supplier: Matrix Scientific
049785-500MG
049785-2.5G
101889-040EA
364.62
USD
101889-040
101889-042
1-{4-[2-(3-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride
1-{4-[2-(3-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₂ClNO₂ |
MDL Number:
MFCD13560187 CAS Number: 1219968-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...