1-{4-[2-(4-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride

Supplier: Matrix Scientific

049784-500MG 049784-2.5G
101889-036EA 364.62 USD
101889-036 101889-038
1-{4-[2-(4-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride
1-{4-[2-(4-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₂
MDL Number: MFCD13560186
CAS Number: 1219971-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR