1-{2-[2-(2-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride

Supplier: Matrix Scientific

049782-500MG 049782-2.5G
101889-028EA 364.62 USD
101889-028 101889-030
1-{2-[2-(2-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride
1-{2-[2-(2-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₂
MDL Number: MFCD13560184
CAS Number: 1220028-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR