1-{2-[2-(4-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride

Supplier: Matrix Scientific

049780-500MG 049780-2.5G
101889-020EA 364.62 USD
101889-020 101889-022
1-{2-[2-(4-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride
1-{2-[2-(4-Piperidinyl)ethoxy]phenyl}-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₂
MDL Number: MFCD13560182
CAS Number: 1220031-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR