5-Chloro-8-(3-pyrrolidinylmethoxy)quinoline hydrochloride

Supplier: Matrix Scientific

049779-500MG 049779-2.5G
101889-016EA 364.62 USD
101889-016 101889-018
5-Chloro-8-(3-pyrrolidinylmethoxy)quinoline hydrochloride
5-Chloro-8-(3-pyrrolidinylmethoxy)quinoline hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₆Cl₂N₂O
MDL Number: MFCD13560181
CAS Number: 1219968-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR