5-Chloro-8-quinolinyl-2-(2-piperidinyl)ethylether hydrochloride

Supplier: Matrix Scientific

049778-500MG 049778-2.5G
101889-012EA 364.62 USD
101889-012 101889-014
5-Chloro-8-quinolinyl-2-(2-piperidinyl)ethylether hydrochloride
5-Chloro-8-quinolinyl-2-(2-piperidinyl)ethylether hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₀Cl₂N₂O
MW: 327.26 g/mol
MDL Number: MFCD13560180
CAS Number: 1220031-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR