4-Bromo-2-nitrophenyl-2-(2-piperidinyl)ethylether hydrochloride

Supplier: Matrix Scientific

049774-500MG 049774-2.5G
101888-996EA 364.62 USD
101888-996 101888-998
4-Bromo-2-nitrophenyl-2-(2-piperidinyl)ethylether hydrochloride
4-Bromo-2-nitrophenyl-2-(2-piperidinyl)ethylether hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrClN₂O₃
MW: 365.66 g/mol
MDL Number: MFCD13560176
CAS Number: 1219964-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR