4-Bromo-2-nitrophenyl-2-(4-piperidinyl)ethylether hydrochloride
Supplier: Matrix Scientific
049772-500MG
049772-2.5G
101888-988EA
364.62
USD
101888-988
101888-990
4-Bromo-2-nitrophenyl-2-(4-piperidinyl)ethylether hydrochloride
4-Bromo-2-nitrophenyl-2-(4-piperidinyl)ethylether hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈BrClN₂O₃ MW: 365.66 g/mol |
MDL Number:
MFCD13560174 CAS Number: 1219967-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...