3-[(4-Chloro-2-nitrophenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
049771-500MG
049771-2.5G
101888-984EA
364.62
USD
101888-984
101888-986
3-[(4-Chloro-2-nitrophenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Chloro-2-nitrophenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₄Cl₂N₂O₃ |
MDL Number:
MFCD13560173 CAS Number: 1220031-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...