3-[2-(4-Chloro-2-nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049769-500MG 049769-2.5G
101888-976EA 364.62 USD
101888-976 101888-978
3-[2-(4-Chloro-2-nitrophenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Chloro-2-nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₂N₂O₃
MDL Number: MFCD13560171
CAS Number: 1220034-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR