Ethyl-4-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride
Supplier: Matrix Scientific
049766-500MG
049766-2.5G
101888-964EA
364.62
USD
101888-964
101888-966
Ethyl-4-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride
Ethyl-4-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₄ClNO₃ |
MDL Number:
MFCD13560168 CAS Number: 1219964-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...