Ethyl-4-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

049765-500MG 049765-2.5G
101888-960EA 364.62 USD
101888-960 101888-962
Ethyl-4-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride
Ethyl-4-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13560167
CAS Number: 1220029-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR