Ethyl-4-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Supplier: Matrix Scientific
049764-500MG
049764-2.5G
101888-956EA
364.62
USD
101888-956
101888-958
Ethyl-4-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Ethyl-4-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₄ClNO₃ MW: 313.83 g/mol |
MDL Number:
MFCD13560166 CAS Number: 136125-47-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...