Methyl-2-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

049762-500MG 049762-2.5G
101888-948EA 364.62 USD
101888-948 101888-950
Methyl-2-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride
Methyl-2-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13560164
CAS Number: 1220027-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR