Methyl-2-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

049760-500MG 049760-2.5G
101888-940EA 364.62 USD
101888-940 101888-942
Methyl-2-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Methyl-2-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13560162
CAS Number: 1219964-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR