N,N-Diethyl-3-[2-(2-piperidinyl)ethoxy]aniline hydrochloride

Supplier: Matrix Scientific

049758-500MG 049758-2.5G
101888-932EA 364.62 USD
101888-932 101888-934
N,N-Diethyl-3-[2-(2-piperidinyl)ethoxy]aniline hydrochloride
N,N-Diethyl-3-[2-(2-piperidinyl)ethoxy]aniline hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₉ClN₂O
MDL Number: MFCD13560160
CAS Number: 1220016-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR