N,N-Diethyl-3-[2-(4-piperidinyl)ethoxy]aniline hydrochloride
Supplier: Matrix Scientific
049756-500MG
049756-2.5G
101888-924EA
364.62
USD
101888-924
101888-926
N,N-Diethyl-3-[2-(4-piperidinyl)ethoxy]aniline hydrochloride
N,N-Diethyl-3-[2-(4-piperidinyl)ethoxy]aniline hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₉ClN₂O |
MDL Number:
MFCD13560158 CAS Number: 1219967-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...