2-{2-[2-(tert-Butyl)-4-methylphenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049754-500MG 049754-2.5G
101888-916EA 364.62 USD
101888-916 101888-918
2-{2-[2-(tert-Butyl)-4-methylphenoxy]ethyl}piperidine hydrochloride
2-{2-[2-(tert-Butyl)-4-methylphenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₃₀ClNO
MDL Number: MFCD13560156
CAS Number: 1219956-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR