3-{2-[2-(tert-Butyl)-4-methylphenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049753-500MG 049753-2.5G
101888-912EA 364.62 USD
101888-912 101888-914
3-{2-[2-(tert-Butyl)-4-methylphenoxy]ethyl}piperidine hydrochloride
3-{2-[2-(tert-Butyl)-4-methylphenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₃₀ClNO
MDL Number: MFCD13560155
CAS Number: 1219964-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR